Learn More
Research Products International Corp SLIDE-OUT FREEZER RACK FOR 2"" BOXES 20 BOX CAPACITY 5X4 ARRAY, 22-1/8 X 5-1/2 X 12""
SDP
Catalog No. 50998150
Encompass
Ideal freezer racks feature slide out trays that maximize storage space in upright freezers or refrigerators. Constructed of corrosion resistant stainless steel for rugged durability. Racks will accommodate all 5" square x 2" high cardboard or plastic boxes. Racks are available in several convenient sizes and capacities.
Catalog No. 50-998-150 Supplier Research Products International Corp Supplier No. 247474
Ideal freezer racks feature slide out trays that maximize storage space in upright freezers or refrigerators. Constructed of corrosion resistant stainless steel for rugged durability. Racks will accommodate all 5" square x 2" high cardboard or plastic boxes. Racks are available in several convenient sizes and capacities.