Learn More
Leica Biosystems SLIDE PINK SNOWCOAT CLIPPED CO
Supplier:  Leica Biosystems 3800386
Encompass_Preferred
Snowcoat Clipped Corner Slides are clipped at 45° angles to help reduce glass breakage and fragmentation when loaded into slide printers.These slides have been optimized for use with the Leica printer.
Catalog No. 50-255-2445