Learn More
Med Vet International McKesson Premium Plain Microscope Slide, 25 x 75 mm, 70-101PMCK, 72/Box
Catalog No. 503043110
Encompass_Preferred
Microscope Slide McKesson 25 X 75 X 1 mm Plain 25 x 75 x 1 millimeters Ground edges Made from the finest quality sheet glass Guaranteed precleaned Non-corrosive glass Fully annealed
Catalog No. 50-304-3110 Supplier Med Vet International Supplier No. 938360BX
Microscope Slide McKesson 25 X 75 X 1 mm Plain 25 x 75 x 1 millimeters Ground edges Made from the finest quality sheet glass Guaranteed precleaned Non-corrosive glass Fully annealed