Learn More
Electron Microscopy Sciences Stainless Steel Slide Rack Adapter
SDP
Catalog No. 501928712
Encompass_Preferred
The Stainless Slide Rack Adapter enables the user to stain both Mega and Supa Mega slides on automated staining equipment, bringing convenience, efficiency, and cost saving all at the same time. This makes the slide rack adapter ideal for all laboratories looking to improve their processes. Stainless steel for a lifetime of use. Enables a LEAN staining process. Use with Mega, Supa Mega, and potentially any bespoke sized slide.
Catalog No. 50-192-8712 Supplier Electron Microscopy Sciences Supplier No. 7006530
The Stainless Slide Rack Adapter enables the user to stain both Mega and Supa Mega slides on automated staining equipment, bringing convenience, efficiency, and cost saving all at the same time. This makes the slide rack adapter ideal for all laboratories looking to improve their processes. Stainless steel for a lifetime of use. Enables a LEAN staining process. Use with Mega, Supa Mega, and potentially any bespoke sized slide.