Learn More
Electron Microscopy Sciences 30-Slide Stainless Steel Rack 1 each
SDP
Catalog No. 5028697
Encompass_Preferred
This 30-slide rack is made from non-tarnishing stainless steel and it is resistant to most staining solutions. The handle is permanently attached but it is swing-able to permitting for closure of the dish and easy insertion and removal of the slides.
Catalog No. 50-286-97 Supplier Electron Microscopy Sciences Supplier No. 7031234
This 30-slide rack is made from non-tarnishing stainless steel and it is resistant to most staining solutions. The handle is permanently attached but it is swing-able to permitting for closure of the dish and easy insertion and removal of the slides.