Learn More
LabScientific Freezer storage rack for 100-place slide boxes. 3x7 compartments (21).
Catalog No. NC1095827
Encompass
Freezer storage rack for 100-place slide boxes. 3x7 compartments (21). Compartment dimensions: l x h x w: 7 3/16 x 1 1/2 x 8 5/16 inch. Exterior dimensions: L x H x W : 22 1/16 x 10 7/8 x 8 3/8 inch
Catalog No. NC1095827 Supplier LabScientific Supplier No. USB37100S
Freezer storage rack for 100-place slide boxes. 3x7 compartments (21). Compartment dimensions: l x h x w: 7 3/16 x 1 1/2 x 8 5/16 inch. Exterior dimensions: L x H x W : 22 1/16 x 10 7/8 x 8 3/8 inch