Learn More
Fine Science Tools Slide Retractor - 2 x 2 Blunt, 2 cm
SDP
Catalog No. NC1325756
Encompass
Product: Slide Retractor - 2x2 Blunt/2cm/1cm Maximum Spread/ Teeth: 2x2 Blunt/ Teeth Length: 4mm/ Teeth Width: 3mm/ Maximum Spread: 1cm/ Alloy: Stainless Steel/ Length: 2cm/Unit: Each
Catalog No. NC1325756 Supplier Fine Science Tools Supplier No. 1700503
Product: Slide Retractor - 2x2 Blunt/2cm/1cm Maximum Spread/ Teeth: 2x2 Blunt/ Teeth Length: 4mm/ Teeth Width: 3mm/ Maximum Spread: 1cm/ Alloy: Stainless Steel/ Length: 2cm/Unit: Each