Learn More
Fine Science Tools SLIDE RETRACTOR - ATRAUMATIC
SDP
Catalog No. NC1325753
Encompass
Product: Slide Retractor - Atraumatic Blades/2cm/1cm Maximum Spread/ Teeth: Atraumatic Blades/ Teeth Length: 4mm/ Teeth Width: 3mm/ Maximum Spread: 1cm/ Alloy: Stainless Steel/ Length: 2cm/Unit: Each
Catalog No. NC1325753 Supplier Fine Science Tools Supplier No. 1700403
Product: Slide Retractor - Atraumatic Blades/2cm/1cm Maximum Spread/ Teeth: Atraumatic Blades/ Teeth Length: 4mm/ Teeth Width: 3mm/ Maximum Spread: 1cm/ Alloy: Stainless Steel/ Length: 2cm/Unit: Each