Learn More
Ted Pella Inc Slide-Sette Saver, Natural, 25/pk
SDP
Catalog No. NC1750752
Encompass
The Slide-Sette Saver offers a convenient single container for shipping a paraffin tissue block together with its corresponding slide. The sturdy molded polypropylene container nestles the paraffin tissue block securely while offering a safe residence for its companion slide.
Catalog No. NC1750752 Supplier Ted Pella Inc Supplier No. 22524NAT
The Slide-Sette Saver offers a convenient single container for shipping a paraffin tissue block together with its corresponding slide. The sturdy molded polypropylene container nestles the paraffin tissue block securely while offering a safe residence for its companion slide.