Learn More
Andwin Scientific SLIDE STAIN RACK S.S
SDP
Catalog No. NC9572056
Encompass_Preferred
The Micro Slide Staining Rack is made of stainless steel. This rack can be easily and rapidly modified in order to fit in sinks up to 53.4 cm (21") inside dimensions. This product is sold individually.
Catalog No. NC9572056 Supplier Andwin Scientific Supplier No. 15153821
The Micro Slide Staining Rack is made of stainless steel. This rack can be easily and rapidly modified in order to fit in sinks up to 53.4 cm (21") inside dimensions. This product is sold individually.