Learn More
Andwin Scientific SLIDE STAINING JAR 2/CS
SDP
Catalog No. 50931013
Encompass_Preferred
The EasyDip Stainless Steel Holder is specially made to hold six EasyDip staining jars together. This product measures 16"L x 4"W x 1 "H (425 x 102 x 38 mm) and is sold in a case of 2 jars in the color of blue.
Catalog No. 50-931-013 Supplier Andwin Scientific Supplier No. 71385B
The EasyDip Stainless Steel Holder is specially made to hold six EasyDip staining jars together. This product measures 16"L x 4"W x 1 "H (425 x 102 x 38 mm) and is sold in a case of 2 jars in the color of blue.