Learn More
Electron Microscopy Sciences Easy Dip™ Slide Staining Jar, Pink
SDP
Catalog No. 501899557
Encompass_Preferred
EasyDip Slide Staining system jars resist attacks from most staining reagents but not phenols. Wide base for stability. 2½" x 3" x 3⅝"
Color: Pink
Catalog No. 50-189-9557 Supplier Electron Microscopy Sciences Supplier No. 71385P
EasyDip Slide Staining system jars resist attacks from most staining reagents but not phenols. Wide base for stability. 2½" x 3" x 3⅝"
Color: Pink