Learn More
Electron Microscopy Sciences Easy Dip™ Slide Staining Jar, Yellow
SDP
Catalog No. 501899559
Encompass_Preferred
EasyDip Slide Staining system jars resist attacks from most staining reagents but not phenols. Wide base for stability. 2½" x 3" x 3⅝"
Color: Yellow
Catalog No. 50-189-9559 Supplier Electron Microscopy Sciences Supplier No. 71385Y
EasyDip Slide Staining system jars resist attacks from most staining reagents but not phenols. Wide base for stability. 2½" x 3" x 3⅝"
Color: Yellow