Learn More
Andwin Scientific SLIDE STAINING SET
SDP
Catalog No. NC9479355
Encompass_Preferred
The Tissue-Tek Slide Staining Set is ideal for use in histological or cytological staining. This product offers concurrent utilization of up to 12 solution wells. It also features a snap-locking cover on solvent-resistant metal frame that eliminates the risk of spills. This set contains 10 white polypropylene dishes, 2 green solvent-resistant dishes, 12 lids, 12-unit metal carrier, and drain tray/cover. It is sold individually.
Catalog No. NC9479355 Supplier Andwin Scientific Supplier No. 25608902
The Tissue-Tek Slide Staining Set is ideal for use in histological or cytological staining. This product offers concurrent utilization of up to 12 solution wells. It also features a snap-locking cover on solvent-resistant metal frame that eliminates the risk of spills. This set contains 10 white polypropylene dishes, 2 green solvent-resistant dishes, 12 lids, 12-unit metal carrier, and drain tray/cover. It is sold individually.