Learn More
Medicus Health Slide Staining Set, (12) Autoclavable Dishes with Hinged Lids, Slide Dipper
SDP
Catalog No. NC3386469
Encompass
Slide Staining Set includes: (12) Autoclavable Dishes with Hinged Lids, (1) 12 Place Stainless Steel Dish Holding Rack & (1) Slide Dipper
∙ Great for immunohistochemistry procedures, histological, cytological and hematological staining procedures as well with total immersion for slides
∙ Dishes and Dippers can be sterilized by autoclave, used in the microwave and are resistant to temperature between 0°C and 120°C
∙ Overall Dimensions: 28"W x 5"D x 5"H
∙ Staining Dish is made from polyester, a thermo plastic resin which is great for electrical, chemical resistance, performs to at high temperatures
∙ Dish capacity is 300 mL and dish is resistant to acids and solvents
∙Staining Dish is designed for 75mm x 25mm or 3" x 1" slides; Dish measures: 5"L x 2.25"W x 4.75"H; Slide Dipper measures: 3.75"L x 1.25"W x 3.75"H
Catalog No. NC3386469 Supplier Medicus Health Supplier No. 7352M2
Slide Staining Set includes: (12) Autoclavable Dishes with Hinged Lids, (1) 12 Place Stainless Steel Dish Holding Rack & (1) Slide Dipper
∙ Great for immunohistochemistry procedures, histological, cytological and hematological staining procedures as well with total immersion for slides
∙ Dishes and Dippers can be sterilized by autoclave, used in the microwave and are resistant to temperature between 0°C and 120°C
∙ Overall Dimensions: 28"W x 5"D x 5"H
∙ Staining Dish is made from polyester, a thermo plastic resin which is great for electrical, chemical resistance, performs to at high temperatures
∙ Dish capacity is 300 mL and dish is resistant to acids and solvents
∙Staining Dish is designed for 75mm x 25mm or 3" x 1" slides; Dish measures: 5"L x 2.25"W x 4.75"H; Slide Dipper measures: 3.75"L x 1.25"W x 3.75"H