Learn More
Medicus Health Slide Staining Set, (3) Autoclavable Dishes, Hinged Lids, Slide Dipper
SDP
Catalog No. NC3386468
Encompass
Slide Staining Set includes: (3) Autoclavable Dishes with Attached Hinged Lids, (1) Stainless Steel Dish Holding Rack & (1) Slide Dipper
∙ Great for immunohistochemistry procedures, histological, cytological and hematological staining procedures as well with total immersion for slides
∙ Dishes and Dippers can be sterilized by autoclave, used in the microwave and are resistant to temperature between 0°C and 120°C.
∙ Triple Staining Station great for busy Hematology Departments who need to do a quick Wright's Stain
Compact
∙ Multiple units can be placed in a row to build any specialty staining procedure
∙ Overall Dimensions: 9"W x 5"D x 5"H
∙ Staining Dish is designed for 75mm x 25mm or 3" x 1" slides; Dish measures: 5"L x 2.25"W x 4.75"H;
Slide Dipper measures: 3.75"L x 1.25"W x 3.75"H
Catalog No. NC3386468 Supplier Medicus Health Supplier No. 7352M1
Slide Staining Set includes: (3) Autoclavable Dishes with Attached Hinged Lids, (1) Stainless Steel Dish Holding Rack & (1) Slide Dipper
∙ Great for immunohistochemistry procedures, histological, cytological and hematological staining procedures as well with total immersion for slides
∙ Dishes and Dippers can be sterilized by autoclave, used in the microwave and are resistant to temperature between 0°C and 120°C.
∙ Triple Staining Station great for busy Hematology Departments who need to do a quick Wright's Stain
Compact
∙ Multiple units can be placed in a row to build any specialty staining procedure
∙ Overall Dimensions: 9"W x 5"D x 5"H
∙ Staining Dish is designed for 75mm x 25mm or 3" x 1" slides; Dish measures: 5"L x 2.25"W x 4.75"H;
Slide Dipper measures: 3.75"L x 1.25"W x 3.75"H