Learn More
Medicus Health Slide Staining Station, Three Clear Dishes with Slide Dipper
SDP
Catalog No. NC3448936
Encompass
Slide Staining Set includes: (3) Clear Dishes with Lids, (1) Plastic Slide Dipper and (1) three bank stainless steel dish holding rack
∙ Great for immunohistochemistry procedures, histological, cytological and hematological staining procedures as well with total immersion for slides
∙ Three Place Staining Station great for busy Hematology Departments who need to do H&E staining procedures (Hematoxylin & Eosin)
∙ Overall Stain Set Dimensions: 8.75"W x 5"D x 5"H
∙ Clear dishes are made from hydrocarbon polymers of acetal resin and should be used for xylol or other cyclic-ring solvents
∙ Dish capacity is 300 mL and dish is resistant to acids and solvents
∙Staining Dish is designed for 75mm x 25mm or 3" x 1" slides; Dish measures: 4"L x 2"W x 4"H; Slide Dipper measures: 3.6"L x 1.2"W x 3.6"H
Catalog No. NC3448936 Supplier Medicus Health Supplier No. 2935M15
Slide Staining Set includes: (3) Clear Dishes with Lids, (1) Plastic Slide Dipper and (1) three bank stainless steel dish holding rack
∙ Great for immunohistochemistry procedures, histological, cytological and hematological staining procedures as well with total immersion for slides
∙ Three Place Staining Station great for busy Hematology Departments who need to do H&E staining procedures (Hematoxylin & Eosin)
∙ Overall Stain Set Dimensions: 8.75"W x 5"D x 5"H
∙ Clear dishes are made from hydrocarbon polymers of acetal resin and should be used for xylol or other cyclic-ring solvents
∙ Dish capacity is 300 mL and dish is resistant to acids and solvents
∙Staining Dish is designed for 75mm x 25mm or 3" x 1" slides; Dish measures: 4"L x 2"W x 4"H; Slide Dipper measures: 3.6"L x 1.2"W x 3.6"H