Learn More
Medicus Health Slide Staining Station, 3 Place, 3 Dishes With Lids & Slide Dipper
SDP
Catalog No. NC2014629
Encompass
∙ Slide Staining 3 Place Station, 3 Dishes with Lids and Slide Dipper are great for histological, cytological and hematological staining procedures with total immersion for slides ∙Slide Staining Station has (3) dishes with lids, (1) plastic slide dipper and (1) three bank stainless steel dish holding rack∙Staining Station is designed for 75mm x 25mm or 3" x 1" slides∙Plastic staining dipper holds maximum of 24 slides∙Staining Set Overall Dimensions: 9"W x 5"D x 5"H∙Slide Dipper measures: 3.6"L x 1.2" W x 3.6"H; Each Dish measures: 4"L x 2"W x 4"H
This item is not returnable. View return policy
Catalog No. NC2014629 Supplier Medicus Health Supplier No. 2935M4
∙ Slide Staining 3 Place Station, 3 Dishes with Lids and Slide Dipper are great for histological, cytological and hematological staining procedures with total immersion for slides ∙Slide Staining Station has (3) dishes with lids, (1) plastic slide dipper and (1) three bank stainless steel dish holding rack∙Staining Station is designed for 75mm x 25mm or 3" x 1" slides∙Plastic staining dipper holds maximum of 24 slides∙Staining Set Overall Dimensions: 9"W x 5"D x 5"H∙Slide Dipper measures: 3.6"L x 1.2" W x 3.6"H; Each Dish measures: 4"L x 2"W x 4"H