Learn More
Medicus Health Slide Staining Station, Twelve Clear Dishes with Slide Dipper
SDP
Catalog No. NC2590044
Encompass
∙ Slide Staining Station has (12) clear dishes with lids, (1) plastic slide dipper and (1) twelve bank stainless steel dish holding rack∙ Great for histological, cytological and hematological staining procedures with total immersion for slides∙ Compact, lightweight, durable stainless steel staining rack makes it easy to move and reposition∙ Clear dishes are made from hydrocarbon polymers of acetal resin and should be used for xylol or other cyclic-ring solvents∙ Each Stain Dish has a lid to help reduce reagent evaporation or contamination ∙ Dish capacity is 300 mL and dish is resistant to acids and solvents∙ Use detergent to cleaning dishes∙ Dish is designed for 75mm x 25mm or 3" x 1" slides∙ Plastic staining dipper holds maximum of 24 slides∙ Staining Set Overall Dimensions: 28"W x 5"D x 5"H∙ Slide Dipper measures: 3.6"L x 1.2" W x 3.6"H; Each Dish measures: 4"L x 2"W x 4"H
Catalog No. NC2590044 Supplier Medicus Health Supplier No. 2935M16
∙ Slide Staining Station has (12) clear dishes with lids, (1) plastic slide dipper and (1) twelve bank stainless steel dish holding rack∙ Great for histological, cytological and hematological staining procedures with total immersion for slides∙ Compact, lightweight, durable stainless steel staining rack makes it easy to move and reposition∙ Clear dishes are made from hydrocarbon polymers of acetal resin and should be used for xylol or other cyclic-ring solvents∙ Each Stain Dish has a lid to help reduce reagent evaporation or contamination ∙ Dish capacity is 300 mL and dish is resistant to acids and solvents∙ Use detergent to cleaning dishes∙ Dish is designed for 75mm x 25mm or 3" x 1" slides∙ Plastic staining dipper holds maximum of 24 slides∙ Staining Set Overall Dimensions: 28"W x 5"D x 5"H∙ Slide Dipper measures: 3.6"L x 1.2" W x 3.6"H; Each Dish measures: 4"L x 2"W x 4"H