Learn More
Newcomer Supply StainTray 30 Place with Clear Lid. Holds up to 30 slides on plastic rails covered with a polymer strip to hold slides even at an angle.
SDP
Catalog No. NC2102249
Encompass
The Simport StainTray™ has a black base made of tough ABS plastic withstanding a wide range of chemicals (Avoid chlorinated hydrocarbons ). It will accept up to 30 slides on six plastic rails covered with a red polymer strip to perfectly hold slides even if tray is held at an angle. When humidity is needed, wells between rails will hold up to one ml of water securely without splashing. Middle wells will hold up to 2 ml each. Rails are raised not only to avoid water touching the slides but to make them more easily retrieveable. The base will also hold excess stain solution dripping from the slides. Four rubber feet ensure greater base stability. The lid is Clear to allow for visual examination and is made of PETG with a temp. range of -20°C to +60°C. Units are stackable for space saving purposes. Not autoclavable. Dimensions: 38.4 x 32.9 x 4.5 cm (H).
This item is not returnable. View return policy
Catalog No. NC2102249 Supplier Newcomer Supply Supplier No. 684730CL
The Simport StainTray™ has a black base made of tough ABS plastic withstanding a wide range of chemicals (Avoid chlorinated hydrocarbons ). It will accept up to 30 slides on six plastic rails covered with a red polymer strip to perfectly hold slides even if tray is held at an angle. When humidity is needed, wells between rails will hold up to one ml of water securely without splashing. Middle wells will hold up to 2 ml each. Rails are raised not only to avoid water touching the slides but to make them more easily retrieveable. The base will also hold excess stain solution dripping from the slides. Four rubber feet ensure greater base stability. The lid is Clear to allow for visual examination and is made of PETG with a temp. range of -20°C to +60°C. Units are stackable for space saving purposes. Not autoclavable. Dimensions: 38.4 x 32.9 x 4.5 cm (H).