Learn More
Electron Microscopy Sciences Platinum Line Advanced Adhesive Slides, Straight Edge, 90° Corner, White Label
SDP
Catalog No. 501929071
Encompass_Preferred
Platinum Line Starfrost advanced adhesive microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates. For IHC stainers.
Edge: Straight
Corner: 90°
Label: White
This item is not returnable. View return policy
Catalog No. 50-192-9071 Supplier Electron Microscopy Sciences Supplier No. 71887126
Platinum Line Starfrost advanced adhesive microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates. For IHC stainers.
Edge: Straight
Corner: 90°
Label: White