Learn More
Electron Microscopy Sciences Platinum Line Starfrost Starfrost Slides, Beveled Edge, Clipped Corner, Pink Label
SDP
Catalog No. 501929014
Encompass_Preferred
Platinum Line Starfrost color-coded microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Edge: Beveled
Corner: Clipped
Label: Pink
This item is not returnable. View return policy
Catalog No. 50-192-9014 Supplier Electron Microscopy Sciences Supplier No. 7188767
Platinum Line Starfrost color-coded microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Edge: Beveled
Corner: Clipped
Label: Pink