Learn More
Electron Microscopy Sciences Platinum Line Starfrost Adhesive Slide with 45° Beveled Edge, Yellow
SDP
Supplier:  Electron Microscopy Sciences 71887113
Encompass_Preferred
Platinum Line Starfrost color-coded adhesive microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Color: Yellow
Catalog No. 50-192-9058