Learn More
Electron Microscopy Sciences Platinum Line Starfrost Starfrost Slides, Straight Edge, 90° Corner, Green Label
SDP
Catalog No. 501928989
Encompass_Preferred
Platinum Line Starfrost color-coded microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Edge: Straight
Corner: 90°
Label: Green
This item is not returnable. View return policy
Catalog No. 50-192-8989 Supplier Electron Microscopy Sciences Supplier No. 7188742
Platinum Line Starfrost color-coded microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Edge: Straight
Corner: 90°
Label: Green