Learn More
Electron Microscopy Sciences Platinum Line Starfrost Starfrost Slides, Straight Edge, 90° Corner, Orange Label
SDP
Supplier:  Electron Microscopy Sciences 7188748
Encompass_Preferred
Platinum Line Starfrost color-coded microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Edge: Straight
Corner: 90°
Label: Orange
Catalog No. 50-192-8995