Learn More
Electron Microscopy Sciences Platinum Line Starfrost Adhesive Slide with Straight 90° Edge, Salmon
SDP
Catalog No. 501929042
Encompass_Preferred
Platinum Line Starfrost color-coded adhesive microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Color: Salmon
This item is not returnable. View return policy
Catalog No. 50-192-9042 Supplier Electron Microscopy Sciences Supplier No. 7188797
Platinum Line Starfrost color-coded adhesive microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Color: Salmon