Learn More
Electron Microscopy Sciences Platinum Line Starfrost Etched Slides, White/Black
SDP
Catalog No. 501929088
Encompass_Preferred
Platinum Line Starfrost etched microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Color: White/Black
This item is not returnable. View return policy
Catalog No. 50-192-9088 Supplier Electron Microscopy Sciences Supplier No. 71887143
Platinum Line Starfrost etched microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Color: White/Black