Learn More
Electron Microscopy Sciences Platinum Line Starfrost Microscope Slides, Straight Edge, 1.2 mm thick, White Label
SDP
Catalog No. 501929077
Encompass_Preferred
Platinum Line Starfrost microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Edge: Straight
Thickness: 1.2 mm
Label: White
This item is not returnable. View return policy
Catalog No. 50-192-9077 Supplier Electron Microscopy Sciences Supplier No. 71887132
Platinum Line Starfrost microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Edge: Straight
Thickness: 1.2 mm
Label: White