Learn More
Electron Microscopy Sciences Platinum Line Advanced Adhesive Slides, Straight Edge, 90° Corner, White Label
SDP
Supplier:  Electron Microscopy Sciences 71887125
Encompass_Preferred
Platinum Line Starfrost advanced adhesive microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates. For IHC stainers.
Edge: Straight
Corner: 90°
Label: White
Catalog No. 50-192-9070