Learn More
Electron Microscopy Sciences Platinum Line Starfrost Starfrost Slides, Beveled Edge, Clipped Corner, Yellow Label
SDP
Supplier:  Electron Microscopy Sciences 7188771
Encompass_Preferred
Platinum Line Starfrost color-coded microscope slides are guaranteed to have exceptional clarity and cleanliness. They are packaged in thick foil to protect from humidity even in very warm climates.
Edge: Beveled
Corner: Clipped
Label: Yellow
Catalog No. 50-192-9018