Learn More
Medicus Health Slide Staining Station Dish with Lid, Clear
SDP
Catalog No. NC2590050
Encompass
∙ Clear Slide Staining Station Dish with Lid is great for histological, cytological and hematological staining procedures with total immersion for slides∙ Great for hematology departments who need to do Wright's Stains∙ Clear Dish with Clear Lid are sold together∙ Multiple units can be placed in a row to build any specialty staining procedure∙ Clear dishes are made from polypropylene which can be used for xylol or other cyclic-ring solvents∙ Stain Dish with lids to help reduce reagent evaporation or contamination ∙ Dish capacity is 300 mL and dish is resistant to acids and solvents∙ It is recommended that a detergent wash be used in cleaning the dishes∙ Staining Dish is designed for 75mm x 25mm or 3" x 1" slides∙ Dish is non stackable and is not graduated∙ Dish measures: 4"L x 2"W x 4"H∙ Color is clear
Catalog No. NC2590050 Supplier Medicus Health Supplier No. 2935M14
∙ Clear Slide Staining Station Dish with Lid is great for histological, cytological and hematological staining procedures with total immersion for slides∙ Great for hematology departments who need to do Wright's Stains∙ Clear Dish with Clear Lid are sold together∙ Multiple units can be placed in a row to build any specialty staining procedure∙ Clear dishes are made from polypropylene which can be used for xylol or other cyclic-ring solvents∙ Stain Dish with lids to help reduce reagent evaporation or contamination ∙ Dish capacity is 300 mL and dish is resistant to acids and solvents∙ It is recommended that a detergent wash be used in cleaning the dishes∙ Staining Dish is designed for 75mm x 25mm or 3" x 1" slides∙ Dish is non stackable and is not graduated∙ Dish measures: 4"L x 2"W x 4"H∙ Color is clear