Learn More
Ted Pella Inc SLIDE STORAGE BOX 100LIDE BLK
SDP
Supplier:  Ted Pella Inc 21026
Encompass
The Handy Slide Box has capacity of 100 3 in. x 1 in. or 75mm x 25mm slides, numbered index in hinged lid. Soft cork layer in bottom part. Stackable. This large capacity microscope slide box is available in 6 colors. Black is recommended for light sensitive specimens. Made of polystyrene. Outer dimensions: 21 x 17 x 3cm (8.3 in. x 6.7 in. x 1.18 in.)
Catalog No. NC9636540