Learn More
Andwin Scientific SLIDE STORAGE BOX W/CLEAR HIN
SDP
Catalog No. 50819932
Encompass_Preferred
The slide storage box provides a secure verticle storage for slides. The design of this product permits convenient identification and reading of the barcode of the slides. There are 100 numbered slots that can each hold one or two slides. The tray can be removed in order to hold the maximum quantity of 400 slides. This product has a dimension of 3 1/4" W x 9 5/8" D x 3 3/8" H.
Catalog No. 50-819-932 Supplier Andwin Scientific Supplier No. 23247101
The slide storage box provides a secure verticle storage for slides. The design of this product permits convenient identification and reading of the barcode of the slides. There are 100 numbered slots that can each hold one or two slides. The tray can be removed in order to hold the maximum quantity of 400 slides. This product has a dimension of 3 1/4" W x 9 5/8" D x 3 3/8" H.