Learn More
Mopec Slide Transfer Body Board
SDP
Catalog No. 22444080 Shop All Mopec Products
Change view
Click to view available options
Material:
HDPE
Catalog No. Material
22-444-080 HDPE
1 options
Catalog No. 22-444-080 Supplier Mopec Supplier No. FD010
Provides a stable transfer by using high density polyethylene. Mopec™ Slide Transfer Body Board is ideal for cadaver transfer between multi-level work surfaces such as storage racks or mortuary refrigerators.
  • High density polyethylene
  • Perimeter handles offer safe alternative for moving cadavers from cooler to table
  • Wash down with standard Mopec disinfectant
Warranty and Services
1 Year Warranty
Specifications
Product Type Body Tray
Material HDPE
Depth (English) 0.5 in.
Depth (Metric) 1.27 cm