Learn More
Electron Microscopy Sciences Slide Tray
SDP
Catalog No. 50931134
Encompass_Preferred
White, thermo-formed Polystyrene. Holds 20 of 1"x3" standard slides in a horizontal position. A white background makes it easy to distinguish dyes and stains. The tray features a thumb cut to simplify the removal of slides. Measures: 7-1/2"x13 1/2" (17x34 mm)
Catalog No. 50-931-134 Supplier Electron Microscopy Sciences Supplier No. 71500
White, thermo-formed Polystyrene. Holds 20 of 1"x3" standard slides in a horizontal position. A white background makes it easy to distinguish dyes and stains. The tray features a thumb cut to simplify the removal of slides. Measures: 7-1/2"x13 1/2" (17x34 mm)