Learn More
Electron Microscopy Sciences Slide Tray Cabinet
SDP
Catalog No. 50931148
Encompass_Preferred
The outside is made of white polypropylene, the tray rails are polystyrene, and the vertical sliding door is clear acrylic; this 240-slide capacity cabinet holds twelve microscope slide trays (#71500) horizontally, and it is dust free.
Catalog No. 50-931-148 Supplier Electron Microscopy Sciences Supplier No. 71504
The outside is made of white polypropylene, the tray rails are polystyrene, and the vertical sliding door is clear acrylic; this 240-slide capacity cabinet holds twelve microscope slide trays (#71500) horizontally, and it is dust free.