Learn More
Bel-Art™ Slide Tray Cabinet
SCI_ED
Catalog No. S17170 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Slide Capacity:
240
Catalog No. Slide Capacity
S17170 240
1 options
Catalog No. S17170 Supplier Bel-Art™ Supplier No. H445830000/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Provides a dust-free enclosure.
  • White polypropylene outer shell
  • Polystyrene tray rails
  • Vertically sliding door
  • Stores 240 slides
  • Dimensions: 20 x 36 x 19 in. (508 x 914 x 482.6 mm)
Order Info
Slide trays available separately (Cat. No. S17170T).
Specifications
Type Slide Storage Cabinet
Slide Capacity 240
Device Holds 240 slides
For Use With (Equipment) Microscope
Material Polystyrene
WARNING: Cancer - www.P65Warnings.ca.gov