Learn More
Research Products International Corp Slide Tray, Cardboard, 20 Place, 72/cs
SDP
Catalog No. 501977198
Encompass
Slide Folder holds up to twenty standard microscope slides 75mm x 25mm (3"x1"). Ideal system stores and displays individual slides horizontally. Transparent front panels allows quick and convenient viewing of contents. Slides are easily removed by pressing on one end, as opposite end automatically lifts. Front panels fold behind base for additional space saving purposes. Low profile folders are stackable, take minimum space, and temperature resistant from -80°C to +100 °C.
Catalog No. 50-197-7198 Supplier Research Products International Corp Supplier No. 247821
Slide Folder holds up to twenty standard microscope slides 75mm x 25mm (3"x1"). Ideal system stores and displays individual slides horizontally. Transparent front panels allows quick and convenient viewing of contents. Slides are easily removed by pressing on one end, as opposite end automatically lifts. Front panels fold behind base for additional space saving purposes. Low profile folders are stackable, take minimum space, and temperature resistant from -80°C to +100 °C.