Learn More
Research Products International Corp Slide Tray, White, 10 per Case
SDP
Catalog No. 501367556
Encompass
SlideTray is a convenient microscope slide work tray. Holds, stores and displays twenty 75mm x 25mm (3" x 1") slides vertically. Ideal for a variety of applications requiring slide incubation. Each angled slide rests within recessed well allowing complete submerging. Slides are easily removed and replaced into position. Trays fit and nest inside of each other during storage. When SlideTray is stacked, slides are well protected and do not touch tray above.
Made of high impact polystyrene and withstand temperatures up to +90°C.
Catalog No. 50-136-7556 Supplier Research Products International Corp Supplier No. 247822
SlideTray is a convenient microscope slide work tray. Holds, stores and displays twenty 75mm x 25mm (3" x 1") slides vertically. Ideal for a variety of applications requiring slide incubation. Each angled slide rests within recessed well allowing complete submerging. Slides are easily removed and replaced into position. Trays fit and nest inside of each other during storage. When SlideTray is stacked, slides are well protected and do not touch tray above.
Made of high impact polystyrene and withstand temperatures up to +90°C.