Learn More
Med Vet International McKesson Acrylic Microscope Precision Cell Slide, 10-Chamber, 177-3805, 100/Box
Catalog No. 503044103
Encompass_Preferred
Urinalysis Microscope Slide McKesson Plain Produced from optically clear acrylic Ten chambers, each holding a standard volume of 6.3 uL Within each chamber there are 18 circles that are all together visible at 100x magnification These premeasured circles contain specific volumes of urinary sediment within its circumference that allows for the easy calculation of cellular elements present in 1 mL of urine
Catalog No. 50-304-4103 Supplier Med Vet International Supplier No. 1179435BX
Urinalysis Microscope Slide McKesson Plain Produced from optically clear acrylic Ten chambers, each holding a standard volume of 6.3 uL Within each chamber there are 18 circles that are all together visible at 100x magnification These premeasured circles contain specific volumes of urinary sediment within its circumference that allows for the easy calculation of cellular elements present in 1 mL of urine