Learn More
Ibidi USA Slide VI 0.5, Glass Bottom - #1.5H Glass Bottom Coverslip
SDP
Catalog No. NC1395718
Encompass
The ibidi product family is comprised of a variety of ¿-Slides and ¿-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The glass bottom versions of the ¿-Slides and ¿-Dishes are especially designed for TIRF, super resolution, and single-molecule applications. The ¿-Slide VI 0.5 Glass Bottom allows you to perform high resolution microscopy in a volume-saving channel format. The convenient six channel format of the ¿-Slide VI 0.5 Glass Bottom is ideal for static cell cultivation and the application of standard immunofluorescence protocols, like treatment, staining, and microscopy of living or fixed cells. Alternatively, the ¿-Slide VI 0.5 Glass Bottom can be connected to a pump and enables you to observe cells under flow conditions. 6 x 500 ¿m high channels, #1.5H D 263 M Schott glass coverslip, uncoated, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. NC1395718 Supplier Ibidi USA Supplier No. 80607
The ibidi product family is comprised of a variety of ¿-Slides and ¿-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The glass bottom versions of the ¿-Slides and ¿-Dishes are especially designed for TIRF, super resolution, and single-molecule applications. The ¿-Slide VI 0.5 Glass Bottom allows you to perform high resolution microscopy in a volume-saving channel format. The convenient six channel format of the ¿-Slide VI 0.5 Glass Bottom is ideal for static cell cultivation and the application of standard immunofluorescence protocols, like treatment, staining, and microscopy of living or fixed cells. Alternatively, the ¿-Slide VI 0.5 Glass Bottom can be connected to a pump and enables you to observe cells under flow conditions. 6 x 500 ¿m high channels, #1.5H D 263 M Schott glass coverslip, uncoated, sterilized, 15 pcs./box