Learn More
LabScientific Step Up Slide Warmer 220V; 230V; (Outside of US and Canada); Surface holds about 40 slides. Heats up to 75 C. LED digital display.
Catalog No. NC0568931
Encompass
Surface holds about 40 slides. Heats up to 75 C. Clean, dry and ventilated. Stable work surface. Relative humidity:≤80%. Setting time range 1-240min. Dimensions 14 x 14 x 4 in. (35 x 35 x 10cm.). Ship weight 22 pounds.
Catalog No. NC0568931 Supplier LabScientific Supplier No. XH2002230V
Surface holds about 40 slides. Heats up to 75 C. Clean, dry and ventilated. Stable work surface. Relative humidity:≤80%. Setting time range 1-240min. Dimensions 14 x 14 x 4 in. (35 x 35 x 10cm.). Ship weight 22 pounds.