Learn More
Electron Microscopy Sciences Slide Warmer for 23 slides, 220V
SDP
Catalog No. 50364441
Encompass_Preferred
These slide warmers are ideal for use in cytology, histology, pathology and biology for paraffin tissue section mounting. Available in two sizes. Features thermostatic heat control, anodized black surface provides contrast, and LED temperature display, both set temperature and display temperature. Thermostat setting range from room temperature to 70 degree C.
**Note: European Voltage. For Use Within USA, Please see FCAT# 50930986, for 110VFootprint: 10" x 7"
Catalog No. 50-364-441 Supplier Electron Microscopy Sciences Supplier No. 71319220
These slide warmers are ideal for use in cytology, histology, pathology and biology for paraffin tissue section mounting. Available in two sizes. Features thermostatic heat control, anodized black surface provides contrast, and LED temperature display, both set temperature and display temperature. Thermostat setting range from room temperature to 70 degree C.
**Note: European Voltage. For Use Within USA, Please see FCAT# 50930986, for 110VFootprint: 10" x 7"