Learn More
Newcomer Supply Slide Warmer, 10" x 7" Surface with digitial controller
SDP
Catalog No. NC1867292
Encompass
Slide Warmer is ideal for use in histology, cytology, pathology and biology for drying & mounting paraffin tissue sections on slides. Features include: easy to adjust and use, thermal heater ensures even heat transfer, LED Temperature Display, Temperature setting from room temp to 75°C (+/- 2°C). Available in 2 sizes to fit most needs. Approximate capacity is 23 slides, weight is 10 lbs. and power requirements 100W. CE certified and cord is CSA & UL listed.
Catalog No. NC1867292 Supplier Newcomer Supply Supplier No. 6258
Slide Warmer is ideal for use in histology, cytology, pathology and biology for drying & mounting paraffin tissue sections on slides. Features include: easy to adjust and use, thermal heater ensures even heat transfer, LED Temperature Display, Temperature setting from room temp to 75°C (+/- 2°C). Available in 2 sizes to fit most needs. Approximate capacity is 23 slides, weight is 10 lbs. and power requirements 100W. CE certified and cord is CSA & UL listed.