Learn More
Andwin Scientific SLIDE WARMER 66 SLIDES
SDP
Catalog No. NC9793370
Encompass_Preferred
The Slide Warmers are recommended for use in cytology, histology, pathology and biology for paraffin tissue section mounting. This product features thermostatic heat control from room temperature to 70C, anodized black surface provides contrast, and LED temperature display, both set temperature and display temperature. This product can warm 66 slides. It measures 25 x 8 (63x20cm).
Catalog No. NC9793370 Supplier Andwin Scientific Supplier No. 7132010
The Slide Warmers are recommended for use in cytology, histology, pathology and biology for paraffin tissue section mounting. This product features thermostatic heat control from room temperature to 70C, anodized black surface provides contrast, and LED temperature display, both set temperature and display temperature. This product can warm 66 slides. It measures 25 x 8 (63x20cm).