Learn More
Medicus Health Slide Warmer with Hinged Cover, Large
SDP
Catalog No. NC2826999
Encompass
This Slide Warmer is ideal for use in cytology, histology, pathology, and biology for paraffin tissue section mounting. Anodized black surface provides contrast and even heat transfer. Clear plastic lid allows you to clearly see your specimens and add humidity during your heating. Thermostat setting is easy to use and can be set from room temperature to 75° C
Catalog No. NC2826999 Supplier Medicus Health Supplier No. 5311M1
This Slide Warmer is ideal for use in cytology, histology, pathology, and biology for paraffin tissue section mounting. Anodized black surface provides contrast and even heat transfer. Clear plastic lid allows you to clearly see your specimens and add humidity during your heating. Thermostat setting is easy to use and can be set from room temperature to 75° C