Learn More
Electron Microscopy Sciences Microscope Slide Warmer Mh6616, 230 V, 15.5 x 7.12 x 3.5 cm (39 x 18 x 9 in)
SDP
Catalog No. 501899553
Encompass_Preferred
Large size economical slide warmer designed for cytology, histology, pathology, and other biological applications in industrial and clinical lab environments. Temperature range from slightly above ambient to 65 degree C. Large slide capacity with space-saving design. 2 year warranty and CE certified. Uniform heat transfer over entire heating surface. Black Anodized heavy gauge aluminum worktop for maximum specimen contrast. Temperature range to 100ºC with slides across or angled on the drying bars, or lying flat without bars. Uniform heat transfer.
DimensionsW" x D" X H" (cm): 15.5 x 7.12 x 3.5 cm (39 x 18 x 9 in)
Capacity: 50 slides 1" x 3"
Catalog No. 50-189-9553 Supplier Electron Microscopy Sciences Supplier No. 7131720
Large size economical slide warmer designed for cytology, histology, pathology, and other biological applications in industrial and clinical lab environments. Temperature range from slightly above ambient to 65 degree C. Large slide capacity with space-saving design. 2 year warranty and CE certified. Uniform heat transfer over entire heating surface. Black Anodized heavy gauge aluminum worktop for maximum specimen contrast. Temperature range to 100ºC with slides across or angled on the drying bars, or lying flat without bars. Uniform heat transfer.
DimensionsW" x D" X H" (cm): 15.5 x 7.12 x 3.5 cm (39 x 18 x 9 in)
Capacity: 50 slides 1" x 3"