Learn More
Electron Microscopy Sciences Microscope Slide Warmer Mh6616x1, 120 V, 15.5 x 7.12 x 3.5 cm (39 x 18 x 9 in)
SDP
Catalog No. 50930984
Encompass_Preferred
Large size economical slide warmer designed for cytology, histology, pathology, and other biological applications in industrial and clinical lab environments. Temperature range from slightly above ambient to 65 degree C. Large slide capacity with space-saving design. 2 year warranty and CE certified. Uniform heat transfer over entire heating surface. Black Anodized heavy gauge aluminum worktop for maximum specimen contrast. Temperature range to 65ºC. Black anodized heavy gauge aluminum worktop.
DimensionsW" x D" X H" (cm): 15.5 x 7.12 x 3.5 cm (39 x 18 x 9 in)
Capacity: 50 slides 1" x 3"
Catalog No. 50-930-984 Supplier Electron Microscopy Sciences Supplier No. 7131710
Large size economical slide warmer designed for cytology, histology, pathology, and other biological applications in industrial and clinical lab environments. Temperature range from slightly above ambient to 65 degree C. Large slide capacity with space-saving design. 2 year warranty and CE certified. Uniform heat transfer over entire heating surface. Black Anodized heavy gauge aluminum worktop for maximum specimen contrast. Temperature range to 65ºC. Black anodized heavy gauge aluminum worktop.
DimensionsW" x D" X H" (cm): 15.5 x 7.12 x 3.5 cm (39 x 18 x 9 in)
Capacity: 50 slides 1" x 3"