Learn More
Electron Microscopy Sciences Portable Slide Warmer 220 Volt
SDP
Catalog No. 501926589
Encompass_Preferred
With precision temperature control this rugged portable warmer is perfect for many applications.
Its design allows for slides, coverslips and glassware to keep them at the optimum temperature for motility testing. The aluminum heating surface measures 7"" and 11"". The unit features a simple solid state temperature control that is calibrated at 37 degree C (98 degree F) and it is housed in a durable plastic case with a great carrying handle. The hinged lid may be closed when not in use and the power cord is detachable and it stores in the box.
This item is not returnable. View return policy
Catalog No. 50-192-6589 Supplier Electron Microscopy Sciences Supplier No. 7131802
With precision temperature control this rugged portable warmer is perfect for many applications.
Its design allows for slides, coverslips and glassware to keep them at the optimum temperature for motility testing. The aluminum heating surface measures 7"" and 11"". The unit features a simple solid state temperature control that is calibrated at 37 degree C (98 degree F) and it is housed in a durable plastic case with a great carrying handle. The hinged lid may be closed when not in use and the power cord is detachable and it stores in the box.