Learn More
Electron Microscopy Sciences Slide Warmer for 66 slides, 120 V
SDP
Catalog No. 501899554
Encompass_Preferred
These slide warmers are ideal for use in cytology, histology, pathology and biology for paraffin tissue section mounting. Available in two sizes. Features thermostatic heat control, anodized black surface provides contrast, and LED temperature display, both set temperature and display temperature. Thermostat setting range from room temperature to 70 degree C.
Footprint: 25" x 8"
Catalog No. 50-189-9554 Supplier Electron Microscopy Sciences Supplier No. 7132010
These slide warmers are ideal for use in cytology, histology, pathology and biology for paraffin tissue section mounting. Available in two sizes. Features thermostatic heat control, anodized black surface provides contrast, and LED temperature display, both set temperature and display temperature. Thermostat setting range from room temperature to 70 degree C.
Footprint: 25" x 8"